ENAH5802F3A4 Enamine

1H,2H,3H,4H,5H,6H,7H,8H-pyrido[3,4-d]pyrimidine-2,4-dione hydrochloride

  • MDL number MFCD12028208
  • Cas Number : 1185304-70-4
  • Molecular Weight : 203.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427115
Availability details for
Quantity
  • Availability
  • Add to Cart