ENAH5802F3B6 Enamine

1,3-dihydrospiro[indene-2,3'-pyrrolidine] hydrochloride

  • MDL number MFCD02179117
  • Cas Number : 2097996-29-5
  • Molecular Weight : 209.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1215803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1215803
Availability details for
Quantity
  • Availability
  • Add to Cart