ENAH5802F400 Enamine

6'-methyl-1',2'-dihydrospiro[azetidine-3,4'-[3,1]benzoxazine]-2'-one; trifluoroacetic acid

  • MDL number MFCD32066587
  • Cas Number : 2361643-55-0
  • Molecular Weight : 318.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432673
Availability details for
Quantity
  • Availability
  • Add to Cart