ENAH5802F413 Enamine

5-(benzyloxy)-N-methylpyrimidin-2-amine

  • MDL number MFCD24621427
  • Cas Number : 1400807-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427453
Availability details for
Quantity
  • Availability
  • Add to Cart