ENAH5802F416 Enamine

1-methyl-7-(trifluoromethyl)-1,2,3,4-tetrahydroquinoxaline hydrochloride

  • Cas Number : 2344678-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426229
Availability details for
Quantity
  • Availability
  • Add to Cart