ENAH5802F41B Enamine

5-ethyl-3-{2H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridin-7-yl}-1H-1,2,4-triazole dihydrochloride

  • MDL number MFCD32197678
  • Cas Number : 2375270-38-3
  • Molecular Weight : 291.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-(5-ethyl-1H-1,2,4-triazol-3-yl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine dihydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7440075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440075
Availability details for
Quantity
  • Availability
  • Add to Cart