ENAH5802F41E Enamine

methyl 3',4'-dihydro-2'H-spiro[cyclobutane-1,1'-pyrrolo[1,2-a]pyrazine]-6'-carboxylate; trifluoroacetic acid

  • Cas Number : 2375261-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439081
Availability details for
Quantity
  • Availability
  • Add to Cart