ENAH5802F422 Enamine

8-fluoro-1,2-dihydrospiro[3,1-benzoxazine-4,3'-pyrrolidin]-2-one; trifluoroacetic acid

  • MDL number MFCD32066588
  • Cas Number : 2361645-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432678
Availability details for
Quantity
  • Availability
  • Add to Cart