ENAH5802F425 Enamine

{[2-(4-fluorophenyl)-1,3-thiazol-4-yl]methyl}(methyl)amine

  • MDL number MFCD12144168
  • Cas Number : 1153070-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2307146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2307146
Availability details for
Quantity
  • Availability
  • Add to Cart