ENAH5802F42A Enamine

4-(4-phenyl-1H-pyrazol-1-yl)piperidine

  • MDL number MFCD08060998
  • Cas Number : 902836-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5521883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5521883
Availability details for
Quantity
  • Availability
  • Add to Cart