ENAH5802F430 Enamine

2-methyl-4-(trifluoromethyl)-1H,2H,5H,6H,7H,8H-pyrido[3,4-d]pyridazin-1-one hydrochloride

  • MDL number MFCD32068671
  • Cas Number : 2361872-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434728
Availability details for
Quantity
  • Availability
  • Add to Cart