ENAH5802F431 Enamine

6-methoxy-1,2-dihydrospiro[3,1-benzoxazine-4,3'-pyrrolidin]-2-one hydrochloride

  • MDL number MFCD32066578
  • Cas Number : 2361644-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432644
Availability details for
Quantity
  • Availability
  • Add to Cart