ENAH5802F435 Enamine

N-({6-azaspiro[3.4]octan-8-yl}methyl)-2,2,2-trifluoroacetamide hydrochloride

  • MDL number MFCD32066226
  • Cas Number : 2361634-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434057

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434057
Availability details for
Quantity
  • Availability
  • Add to Cart