ENAH5802F43C Enamine

4-[(3-fluoro-4-methoxyphenyl)methoxy]piperidine hydrochloride

  • MDL number MFCD32069341
  • Cas Number : 2375258-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434997
Availability details for
Quantity
  • Availability
  • Add to Cart