ENAH5802F445 Enamine

6-chloro-1,2-dihydrospiro[3,1-benzoxazine-4,4'-piperidin]-2-one hydrochloride

  • MDL number MFCD32066582
  • Cas Number : 85732-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432654
Availability details for
Quantity
  • Availability
  • Add to Cart