ENAH5802F446 Enamine

6-(piperazin-1-yl)-8-oxa-3,5-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),3,5,10,12-hexaene

  • MDL number MFCD01882365
  • Cas Number : 380339-27-5
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(1-piperazinyl)[1]benzofuro[3,2-d]pyrimidine
Inchi Code 1S/C14H14N4O/c1-2-4-11-10(3-1)12-13(19-11)14(17-9-16-12)18-7-5-15-6-8-18/h1-4,9,15H,5-8H2
Inchi Key QWQBHCFTLXGCEJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-29471

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-29471
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart