ENAH5802F449 Enamine

(2,2,2-trifluoroethyl)({[4-(trifluoromethyl)phenyl]methyl})amine

  • MDL number MFCD11127864
  • Cas Number : 1019449-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444193
Availability details for
Quantity
  • Availability
  • Add to Cart