ENAH5802F44B Enamine

(1-phenoxypropan-2-yl)(1-phenylpropan-2-yl)amine

  • MDL number MFCD00867154
  • Cas Number : 1590-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426534
Availability details for
Quantity
  • Availability
  • Add to Cart