ENAH5802F44C Enamine

6-methyl-8-[2-(pyrrolidin-1-yl)pyrimidin-4-yl]-2,6-diazaspiro[3.4]octane trihydrochloride

  • Cas Number : 2375270-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7440587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440587
Availability details for
Quantity
  • Availability
  • Add to Cart