ENAH5802F44E Enamine

[2-(3,4-dimethoxyphenyl)ethyl](2-phenylethyl)amine

  • MDL number MFCD08756299
  • Cas Number : 63925-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1703334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1703334
Availability details for
Quantity
  • Availability
  • Add to Cart