ENAH5802F450 Enamine

methyl 2-{[(tert-butoxy)carbonyl]amino}-3-(piperidin-4-yl)propanoate

  • MDL number MFCD32068938
  • Cas Number : 1822461-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6292168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6292168
Availability details for
Quantity
  • Availability
  • Add to Cart