ENAH5802F455 Enamine

8-tert-butyl 3-ethyl 2,8-diazaspiro[4.5]decane-3,8-dicarboxylate

  • MDL number MFCD12198516
  • Cas Number : 203934-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-304082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-304082
Availability details for
Quantity
  • Availability
  • Add to Cart