ENAH5802F456 Enamine

methyl 4-({[(2R)-1-methoxy-1-oxo-3-(pyridin-2-yl)propan-2-yl]amino}methyl)benzoate dihydrochloride

  • Cas Number : 2343964-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7356214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356214
Availability details for
Quantity
  • Availability
  • Add to Cart