ENAH5802F457 Enamine

methyl 4-({[(2R)-1-methoxy-1-oxo-3-(pyridin-2-yl)propan-2-yl]amino}methyl)benzoate

  • MDL number MFCD31924400
  • Cas Number : 2170006-62-7
  • Molecular Weight : 328.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7356199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356199
Availability details for
Quantity
  • Availability
  • Add to Cart