ENAH5802F459 Enamine

({4-[(2-fluorophenyl)methoxy]-3-methoxyphenyl}methyl)[(pyridin-2-yl)methyl]amine

  • MDL number MFCD07408471
  • Cas Number : 880811-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426404
Availability details for
Quantity
  • Availability
  • Add to Cart