ENAH5802F460 Enamine

5-methyl-4H-1,2,4-triazole-3-sulfonamide

  • MDL number MFCD25980340
  • Cas Number : 29982-54-5
  • Molecular Weight : 162.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 213-215
Iupac Name 5-methyl-4H-1,2,4-triazole-3-sulfonamide
Inchi Code 1S/C3H6N4O2S/c1-2-5-3(7-6-2)10(4,8)9/h1H3,(H2,4,8,9)(H,5,6,7)
Inchi Key VWPHTCYAPDVEFD-UHFFFAOYSA-N

Description

Melting Point 213-215
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-276706

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-276706
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart