ENAH5802F464 Enamine

3-methyl-1-(propan-2-yl)-1H-pyrazole-4-sulfonamide

  • MDL number MFCD21667103
  • Cas Number : 1481935-66-3
  • Molecular Weight : 203.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-isopropyl-3-methyl-1H-pyrazole-4-sulfonamide
Inchi Code 1S/C7H13N3O2S/c1-5(2)10-4-7(6(3)9-10)13(8,11)12/h4-5H,1-3H3,(H2,8,11,12)
Inchi Key VREXESIGMQFVSJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-278592

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-278592
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart