ENAH5802F466 Enamine

6-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonamide

  • MDL number MFCD00649196
  • Cas Number : 446268-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7424880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7424880
Availability details for
Quantity
  • Availability
  • Add to Cart