ENAH5802F46D Enamine

4-methyl-6-(trifluoromethyl)pyridine-3-sulfonamide

  • Cas Number : 2344681-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355659
Availability details for
Quantity
  • Availability
  • Add to Cart