ENAH5802F46E Enamine

2,5-dichloro-4-methylthiophene-3-sulfonamide

  • MDL number MFCD32065994
  • Cas Number : 2361643-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433848
Availability details for
Quantity
  • Availability
  • Add to Cart