ENAH5802F46F Enamine

2-fluoropropane-1-sulfonyl chloride

  • MDL number MFCD10000562
  • Cas Number : 1146664-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7340936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7340936
Availability details for
Quantity
  • Availability
  • Add to Cart