ENAH5802F471 Enamine

hex-5-yne-1-sulfonyl chloride

  • MDL number MFCD19200721
  • Cas Number : 67448-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7041828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7041828
Availability details for
Quantity
  • Availability
  • Add to Cart