ENAH5802F472 Enamine

4-methyl-1,2,5-oxadiazole-3-sulfonyl chloride

  • MDL number MFCD19201276
  • Cas Number : 1198084-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1663673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1663673
Availability details for
Quantity
  • Availability
  • Add to Cart