ENAH5802F473 Enamine

(2-oxocyclobutyl)methanesulfonyl chloride

  • Cas Number : 2375272-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7437513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437513
Availability details for
Quantity
  • Availability
  • Add to Cart