ENAH5802F475 Enamine

(pyridin-3-yl)methanesulfonyl chloride hydrochloride

  • MDL number MFCD28167969
  • Cas Number : 191105-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7437052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437052
Availability details for
Quantity
  • Availability
  • Add to Cart