ENAH5802F47C Enamine

2-cyclopropyl-1,3-oxazole-5-sulfonyl chloride

  • Molecular Weight : 207.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-cyclopropyloxazole-5-sulfonyl chloride
Inchi Code 1S/C6H6ClNO3S/c7-12(9,10)5-3-8-6(11-5)4-1-2-4/h3-4H,1-2H2
Inchi Key LMBLDBCWUJGBKO-UHFFFAOYSA-N

Description

Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-299262

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-299262
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart