ENAH5802F47D Enamine

3-(cyclopropylmethyl)cyclobutane-1-sulfonyl chloride

  • MDL number MFCD32066338
  • Cas Number : 2361635-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7430931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430931
Availability details for
Quantity
  • Availability
  • Add to Cart