ENAH5802F47F Enamine

azocane-1-sulfonyl chloride

  • MDL number MFCD11173328
  • Cas Number : 1042803-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-271510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-271510
Availability details for
Quantity
  • Availability
  • Add to Cart