ENAH5802F480 Enamine

2-(chlorosulfonyl)ethyl N,N-dimethylcarbamate

  • MDL number MFCD19200652
  • Cas Number : 1314911-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1601882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1601882
Availability details for
Quantity
  • Availability
  • Add to Cart