ENAH5802F481 Enamine

6-fluoro-1-(propan-2-yl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD16841035
  • Cas Number : 1250572-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-645814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-645814
Availability details for
Quantity
  • Availability
  • Add to Cart