ENAH5802F486 Enamine

quinoline-7-sulfonyl chloride hydrochloride

  • MDL number MFCD32198461
  • Cas Number : 2375267-39-1
  • Molecular Weight : 264.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name quinoline-7-sulfonyl chloride hydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7443653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443653
Availability details for
Quantity
  • Availability
  • Add to Cart