ENAH5802F488 Enamine

3-(chloromethyl)-1-methyl-1H-pyrazole-4-sulfonyl chloride

  • Cas Number : 2344680-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1717523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1717523
Availability details for
Quantity
  • Availability
  • Add to Cart