ENAH5802F489 Enamine

6,6-difluorospiro[3.3]heptane-2-sulfonyl chloride

  • MDL number MFCD32066638
  • Cas Number : 2361633-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432900
Availability details for
Quantity
  • Availability
  • Add to Cart