ENAH5802F48A Enamine

3-(but-3-en-1-yl)benzene-1-sulfonyl chloride

  • Cas Number : 2375270-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7356785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356785
Availability details for
Quantity
  • Availability
  • Add to Cart