ENAH5802F48B Enamine

2-(but-3-en-1-yl)benzene-1-sulfonyl chloride

  • Cas Number : 1870686-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1585334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1585334
Availability details for
Quantity
  • Availability
  • Add to Cart