ENAH5802F48E Enamine

2,1-benzothiazole-3-sulfonyl chloride

  • MDL number MFCD21760310
  • Cas Number : 128852-03-9
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,1-benzisothiazole-3-sulfonyl chloride
Inchi Code 1S/C7H4ClNO2S2/c8-13(10,11)7-5-3-1-2-4-6(5)9-12-7/h1-4H
Inchi Key HRYOQNVCWHDDPC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-251400

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-251400
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart