ENAH5802F48F Enamine

1-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride

  • Cas Number : 2375268-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444224
Availability details for
Quantity
  • Availability
  • Add to Cart