ENAH5802F490 Enamine

4,5-dimethoxy-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD32068820
  • Cas Number : 168902-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-210
Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427501
Availability details for
Quantity
  • Availability
  • Add to Cart