ENAH5802F494 Enamine

iodomethanesulfonyl chloride

  • MDL number MFCD19200093
  • Cas Number : 22354-05-8
  • Molecular Weight : 240.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name iodomethanesulfonyl chloride
Inchi Code 1S/CH2ClIO2S/c2-6(4,5)1-3/h1H2
Inchi Key HLVPPXHUMMLBLD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-4257713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4257713
Availability details for
Quantity
  • Availability
  • Add to Cart