ENAH5802F497 Enamine

2-hydroxy-1-methyl-1H-1,3-benzodiazole-6-sulfonyl chloride

  • MDL number MFCD28638265
  • Cas Number : 1782422-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7436277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436277
Availability details for
Quantity
  • Availability
  • Add to Cart